missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium iodate, 99%, extra pure, reagent, powder, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 419565000

 View more versions of this product

Catalog No. AC419565000

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
Specifications

Specifications

Sodium iodate
99%
White
Fine Crystalline Powder
98.5% min. (Iodometry)
NaIO3
15, 8764
Solubility in water: soluble. Other solubilities: soluble in acetone, insoluble in alcohol
[O-]I(=O)=O.[Na+]
197.89
CHEBI:81708
99%
Glass bottle
560.0°C
7681-55-2
5.0 to 8.0 (5% soln. at 20°C)
500 g
INaO3
MFCD00003533
sodium iodate, natriumjodat german, iodic acid, sodium salt, sodium iodate naio3, iodic acid hio3 , sodium salt, sodium iodate naio3 pentahydrate, unii-u558pcs5z9, hsdb 749, iodic acid, sodium salt, pentahydrate, natriumjodat
WTCBONOLBHEDIL-UHFFFAOYSA-M
sodium;iodate
23675764
197.89
Extra Pure

Chemical Identifiers

7681-55-2
197.89
WTCBONOLBHEDIL-UHFFFAOYSA-M
23675764
sodium;iodate
INaO3
MFCD00003533
sodium iodate, natriumjodat german, iodic acid, sodium salt, sodium iodate naio3, iodic acid hio3 , sodium salt, sodium iodate naio3 pentahydrate, unii-u558pcs5z9, hsdb 749, iodic acid, sodium salt, pentahydrate, natriumjodat
CHEBI:81708
[O-]I(=O)=O.[Na+]
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May intensify fire; oxidizer.
Harmful if swallowed.
May cause an allergic skin reaction.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Take any precaution to avoid mixing with combustibles.
In case of major

GHS Signal Word: Danger

EINECSNumber : 231-672-5

RTECSNumber : NN1400000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only