Learn More
Sodium iodate, 99%, extra pure, reagent, powder, Thermo Scientific Chemicals
$48.90 - $346.45
Chemical Identifiers
CAS | 7681-55-2 |
---|---|
Molecular Formula | INaO3 |
Molecular Weight (g/mol) | 197.89 |
MDL Number | MFCD00003533 |
InChI Key | WTCBONOLBHEDIL-UHFFFAOYSA-M |
Synonym | sodium iodate, natriumjodat german, iodic acid, sodium salt, sodium iodate naio3, iodic acid hio3 , sodium salt, sodium iodate naio3 pentahydrate, unii-u558pcs5z9, hsdb 749, iodic acid, sodium salt, pentahydrate, natriumjodat |
PubChem CID | 23675764 |
ChEBI | CHEBI:81708 |
IUPAC Name | sodium;iodate |
SMILES | [O-]I(=O)=O.[Na+] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC419560050
|
Thermo Scientific Chemicals
419560050 |
5 g | Glass bottle |
Each for $48.90
|
|
|||||
AC419565000
|
Thermo Scientific Chemicals
419565000 |
500 g | Glass bottle |
Each for $346.45
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentChemical Identifiers
7681-55-2 | |
197.89 | |
WTCBONOLBHEDIL-UHFFFAOYSA-M | |
23675764 | |
sodium;iodate |
INaO3 | |
MFCD00003533 | |
sodium iodate, natriumjodat german, iodic acid, sodium salt, sodium iodate naio3, iodic acid hio3 , sodium salt, sodium iodate naio3 pentahydrate, unii-u558pcs5z9, hsdb 749, iodic acid, sodium salt, pentahydrate, natriumjodat | |
CHEBI:81708 | |
[O-]I(=O)=O.[Na+] |
Specifications
560.0°C | |
White | |
Fine Crystalline Powder | |
98.5% min. (Iodometry) | |
NaIO3 | |
15, 8764 | |
Solubility in water: soluble. Other solubilities: soluble in acetone, insoluble in alcohol | |
[O-]I(=O)=O.[Na+] | |
197.89 | |
CHEBI:81708 | |
99% | |
Glass bottle |
7681-55-2 | |
5.0 to 8.0 (5% soln. at 20°C) | |
5 g | |
INaO3 | |
MFCD00003533 | |
sodium iodate, natriumjodat german, iodic acid, sodium salt, sodium iodate naio3, iodic acid hio3 , sodium salt, sodium iodate naio3 pentahydrate, unii-u558pcs5z9, hsdb 749, iodic acid, sodium salt, pentahydrate, natriumjodat | |
WTCBONOLBHEDIL-UHFFFAOYSA-M | |
sodium;iodate | |
23675764 | |
197.89 | |
Extra Pure | |
Sodium iodate, 99% |
Safety and Handling
GHS H Statement
May intensify fire; oxidizer.
Harmful if swallowed.
May cause an allergic skin reaction.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Take any precaution to avoid mixing with combustibles.
In case of major
GHS Signal Word: Danger
EINECSNumber : 231-672-5
RTECSNumber : NN1400000
TSCA : TSCA
RUO – Research Use Only