missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium iodide dihydrate, 99+%, for analysis, Thermo Scientific Chemicals

$82.61 - $341.05

Chemical Identifiers

CAS 13517-06-1
Molecular Formula INa·2H2O
Molecular Weight (g/mol) 185.93
MDL Number MFCD00149211
InChI Key VEEIYHUOLCUPSA-UHFFFAOYSA-M
Synonym sodium iodide dihydrate, acmc-20ajo4, i.na.2h2o, sodium dihydrate iodide, sodium iodide-water 1/1/2, sodium iodide nai ,dihydrate 9ci
PubChem CID 23685997
IUPAC Name sodium;iodide;dihydrate
SMILES O.O.[Na+].[I-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC202891000
SDS missing translation for 'viewDocuments'
Thermo Scientific Chemicals
202891000
100 g Glass bottle
Each for $82.61
 
AC202895000
SDS missing translation for 'viewDocuments'
Thermo Scientific Chemicals
202895000
500 g Glass bottle
Each for $341.05
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13517-06-1
185.93
VEEIYHUOLCUPSA-UHFFFAOYSA-M
23685997
O.O.[Na+].[I-]
INa·2H2O
MFCD00149211
sodium iodide dihydrate, acmc-20ajo4, i.na.2h2o, sodium dihydrate iodide, sodium iodide-water 1/1/2, sodium iodide nai ,dihydrate 9ci
sodium;iodide;dihydrate

Specifications

752°C
White
Adhering Fine Crystals
99% min. (Argentometry)
NaI·2H2O
01,1087; 02,384; 03,267; 04,456; 06,543; 07,338; 14,89
sodium iodide dihydrate, acmc-20ajo4, i.na.2h2o, sodium dihydrate iodide, sodium iodide-water 1/1/2, sodium iodide nai ,dihydrate 9ci
VEEIYHUOLCUPSA-UHFFFAOYSA-M
sodium;iodide;dihydrate
23685997
≥99%
Glass bottle
2.4400g/mL
13517-06-1
6 to 9 (5% aq. soln.)
100 g
INa·2H2O
MFCD00149211
10, 8469
Solubility in water: soluble. Other solubilities: soluble in alcohol,glycerol and acetone,very soluble in nh3
O.O.[Na+].[I-]
185.93
185.93
Analytical
2.44
Sodium iodide dihydrate, For analysis
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Very toxic to aquatic life.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid releas

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only