missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium L-(+)-tartrate dihydrate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A161870I

 View more versions of this product

Catalog No. AAA161870I


Explore available promotions

Description

Description

Sodium L-(+)-tartrate dihydrate is used in buffers for molecular biology and cell culture applications. It is also used as an emulsifier and a binding agent in food products such as jellies, margarine, and sausage casingsand also as a food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium L-(+)-tartrate dihydrate is used in buffers for molecular biology and cell culture applications. It is also used as an emulsifier and a binding agent in food products such as jellies, margarine, and sausage casingsand also as a food additive.

Solubility
It is soluble in water (290 mg/ml at 20°C). Insoluble in alcohol, and organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to strong oxidizing agents.
Specifications

Specifications

Sodium L-(+)-tartrate dihydrate
∼150°C (in -2H2O)
Odorless
MFCD00150035
6121732
unii-dia7c37aow, disodium tartrate dihydrate, disodium l-+-tartrate dihydrate, dia7c37aow, disodium +-tartrate dihydrate, sodium l-+-tartrate dihydrate, butanedioic acid, 2,3-dihydroxy-2r,3r-, disodium salt, dihydrate, disodium tartarate dihydrate, sodium tartrate nf, sodium tartrate dihydrate mi
FGJLAJMGHXGFDE-UDMGOBQRNA-L
(2R,3R)-2,3-dihydroxybutanedioic acid;sodium;dihydrate
131855972
99%
6106-24-7
1.82
C4H8Na2O8
5000 g
14,8684
It is soluble in water (290mg/ml at 20°C). Insoluble in alcohol,and organic solvents.
O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O
230.08
230.08 (194.06 Anhydrous)

Chemical Identifiers

6106-24-7
230.08
FGJLAJMGHXGFDE-UDMGOBQRNA-L
131855972
O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O
C4H8Na2O8
MFCD00150035
unii-dia7c37aow, disodium tartrate dihydrate, disodium l-+-tartrate dihydrate, dia7c37aow, disodium +-tartrate dihydrate, sodium l-+-tartrate dihydrate, butanedioic acid, 2,3-dihydroxy-2r,3r-, disodium salt, dihydrate, disodium tartarate dihydrate, sodium tartrate nf, sodium tartrate dihydrate mi
(2R,3R)-2,3-dihydroxybutanedioic acid;sodium;dihydrate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 212-773-3

RTECSNumber : WW8150000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only