Learn More
Sodium L-(+)-tartrate dihydrate, 99%, Thermo Scientific Chemicals
$86.44 - $971.62
Chemical Identifiers
CAS | 6106-24-7 |
---|---|
Molecular Formula | C4H8Na2O8 |
Molecular Weight (g/mol) | 230.08 |
MDL Number | MFCD00150035 |
InChI Key | FGJLAJMGHXGFDE-UDMGOBQRNA-L |
Synonym | unii-dia7c37aow, disodium tartrate dihydrate, disodium l-+-tartrate dihydrate, dia7c37aow, disodium +-tartrate dihydrate, sodium l-+-tartrate dihydrate, butanedioic acid, 2,3-dihydroxy-2r,3r-, disodium salt, dihydrate, disodium tartarate dihydrate, sodium tartrate nf, sodium tartrate dihydrate mi |
PubChem CID | 131855972 |
IUPAC Name | (2R,3R)-2,3-dihydroxybutanedioic acid;sodium;dihydrate |
SMILES | O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1618730
|
Thermo Scientific Chemicals
A1618730 |
250 g |
Each for $86.44
|
|
|||||
AAA161870B
|
Thermo Scientific Chemicals
A161870B |
1000 g |
Each for $249.04
|
|
|||||
AAA161870I
|
Thermo Scientific Chemicals
A161870I |
5000 g |
Each for $971.62
|
|
|||||
Description
Sodium L-(+)-tartrate dihydrate is used in buffers for molecular biology and cell culture applications. It is also used as an emulsifier and a binding agent in food products such as jellies, margarine, and sausage casingsand also as a food additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium L-(+)-tartrate dihydrate is used in buffers for molecular biology and cell culture applications. It is also used as an emulsifier and a binding agent in food products such as jellies, margarine, and sausage casingsand also as a food additive.
Solubility
It is soluble in water (290 mg/ml at 20°C). Insoluble in alcohol, and organic solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to strong oxidizing agents.
Chemical Identifiers
6106-24-7 | |
230.08 | |
FGJLAJMGHXGFDE-UDMGOBQRNA-L | |
131855972 | |
O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O |
C4H8Na2O8 | |
MFCD00150035 | |
unii-dia7c37aow, disodium tartrate dihydrate, disodium l-+-tartrate dihydrate, dia7c37aow, disodium +-tartrate dihydrate, sodium l-+-tartrate dihydrate, butanedioic acid, 2,3-dihydroxy-2r,3r-, disodium salt, dihydrate, disodium tartarate dihydrate, sodium tartrate nf, sodium tartrate dihydrate mi | |
(2R,3R)-2,3-dihydroxybutanedioic acid;sodium;dihydrate |
Specifications
6106-24-7 | |
1.82 | |
C4H8Na2O8 | |
250 g | |
14,8684 | |
It is soluble in water (290mg/ml at 20°C). Insoluble in alcohol,and organic solvents. | |
O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O | |
230.08 | |
230.08 (194.06 Anhydrous) | |
Sodium L-(+)-tartrate dihydrate |
∼150°C (in -2H2O) | |
Odorless | |
MFCD00150035 | |
6121732 | |
unii-dia7c37aow, disodium tartrate dihydrate, disodium l-+-tartrate dihydrate, dia7c37aow, disodium +-tartrate dihydrate, sodium l-+-tartrate dihydrate, butanedioic acid, 2,3-dihydroxy-2r,3r-, disodium salt, dihydrate, disodium tartarate dihydrate, sodium tartrate nf, sodium tartrate dihydrate mi | |
FGJLAJMGHXGFDE-UDMGOBQRNA-L | |
(2R,3R)-2,3-dihydroxybutanedioic acid;sodium;dihydrate | |
131855972 | |
99% |
Safety and Handling
EINECSNumber : 212-773-3
RTECSNumber : WW8150000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only