missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Laurate 97.0+%, TCI America™
Supplier: TCI America L001625G
Specifications
| Sodium Laurate | |
| White | |
| MFCD00041754 | |
| 3077 | |
| BTURAGWYSMTVOW-UHFFFAOYSA-M | |
| sodium dodecanoate | |
| 2735067 | |
| ≥97.0% (T) |
| 629-25-4 | |
| C12H23NaO2 | |
| 25 g | |
| sodium dodecanoate, sodium laurate, lauric acid sodium salt, dodecanoic acid, sodium salt, lauric acid, sodium salt, dodecanoic acid sodium salt, lauran sodny czech, caswell no. 778a, unii-k146mr5exo, epa pesticide chemical code 079026 | |
| [Na+].CCCCCCCCCCCC([O-])=O | |
| 222.30 | |
| 222.30 | |
| Crystalline Powder |
Chemical Identifiers
| 629-25-4 | |
| 222.30 | |
| BTURAGWYSMTVOW-UHFFFAOYSA-M | |
| 2735067 | |
| [Na+].CCCCCCCCCCCC([O-])=O |
| C12H23NaO2 | |
| MFCD00041754 | |
| sodium dodecanoate, sodium laurate, lauric acid sodium salt, dodecanoic acid, sodium salt, lauric acid, sodium salt, dodecanoic acid sodium salt, lauran sodny czech, caswell no. 778a, unii-k146mr5exo, epa pesticide chemical code 079026 | |
| sodium dodecanoate |
Safety and Handling
EINECSNumber : (2)-0611&(7)-0973
RTECSNumber : OF0700000
TSCA : Yes