missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium methoxide, pure, anhydrous powder, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 173120050

 View more versions of this product

Catalog No. AC173120050

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium methoxide
124-41-4,497-19-8,1310-73-2
White-Yellow
Glass bottle
CH3ONa
5 g
01, 280
15, 8775
Solubility in water: reacts with water. Other solubilities: soluble in methanol and ethanol, insoluble in hydrocarbons(benzene, toluene, xylene
C[O-].[Na+]
54.02
54.02
Pure
total alkalinity (as NaOCH3):> 99%
126.0°C
0.8
CH3NaO
MFCD00012179
1431
01,1091; 02,385; 03,259; 04,457; 05,617; 06,545; 08,463; 11,489; 13,352
sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755
WQDUMFSSJAZKTM-UHFFFAOYSA-N
sodium;methanolate
10942334
≥97% (NaOCH3)
Powder

Chemical Identifiers

124-41-4
54.02
WQDUMFSSJAZKTM-UHFFFAOYSA-N
10942334
C[O-].[Na+]
CH3NaO
MFCD00012179
sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755
sodium;methanolate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Self-heating; may catch fire.
Flammable solid.
Harmful if swallowed.
May be corrosive to metals.
Reacts violently with water.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 204-699-5

RTECSNumber : PC3570000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only