Learn More
Sodium methoxide, pure, anhydrous powder
CAS: 124-41-4 | CH3NaO | 54.02 g/mol
$73.94 - $160.33
Chemical Identifiers
| CAS | 124-41-4 |
|---|---|
| Molecular Formula | CH3NaO |
| Molecular Weight (g/mol) | 54.02 |
| MDL Number | MFCD00012179 |
| InChI Key | WQDUMFSSJAZKTM-UHFFFAOYSA-N |
| Synonym | sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755 |
| PubChem CID | 10942334 |
| IUPAC Name | sodium;methanolate |
| SMILES | C[O-].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC173120050
|
Thermo Scientific Chemicals
173120050 |
5 g | Glass bottle |
Each for $73.94
|
|
||||
|
AC173121000
|
Thermo Scientific Chemicals
173121000 |
100 g | Glass bottle |
Each for $76.56
|
|
||||
|
AC173125000
|
Thermo Scientific Chemicals
173125000 |
500 g | Glass bottle |
Each for $160.33
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 124-41-4 | |
| 54.02 | |
| WQDUMFSSJAZKTM-UHFFFAOYSA-N | |
| 10942334 | |
| C[O-].[Na+] |
| CH3NaO | |
| MFCD00012179 | |
| sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755 | |
| sodium;methanolate |
Specifications
| total alkalinity (as NaOCH3):> 99% | |
| 126.0°C | |
| 0.8 | |
| CH3NaO | |
| MFCD00012179 | |
| 1431 | |
| 01,1091; 02,385; 03,259; 04,457; 05,617; 06,545; 08,463; 11,489; 13,352 | |
| sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755 | |
| WQDUMFSSJAZKTM-UHFFFAOYSA-N | |
| sodium;methanolate | |
| 10942334 | |
| ≥97% (NaOCH3) | |
| Powder |
| 124-41-4,497-19-8,1310-73-2 | |
| White-Yellow | |
| Glass bottle | |
| CH3ONa | |
| 5 g | |
| 01, 280 | |
| 15, 8775 | |
| Solubility in water: reacts with water. Other solubilities: soluble in methanol and ethanol, insoluble in hydrocarbons(benzene, toluene, xylene | |
| C[O-].[Na+] | |
| 54.02 | |
| 54.02 | |
| Pure | |
| Sodium methoxide |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Self-heating; may catch fire.
Flammable solid.
Harmful if swallowed.
May be corrosive to metals.
Reacts violently with water.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 204-699-5
RTECSNumber : PC3570000
TSCA : TSCA
RUO – Research Use Only