missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Myristate 98.0+%, TCI America™
Supplier: TCI America M0483100G
Specifications
Sodium Myristate | |
822-12-8 | |
100 g | |
MFCD00042653 | |
JUQGWKYSEXPRGL-UHFFFAOYSA-M | |
sodium;tetradecanoate | |
23693095 | |
≥98.0% (GC,T) |
White | |
Crystalline Powder | |
C14H27NaO2 | |
Myristic Acid Sodium Salt | |
CCCCCCCCCCCCCC(=O)[O-].[Na+] | |
250.358 | |
250.36 |
Chemical Identifiers
822-12-8 | |
250.358 | |
JUQGWKYSEXPRGL-UHFFFAOYSA-M | |
23693095 | |
CCCCCCCCCCCCCC(=O)[O-].[Na+] |
C14H27NaO2 | |
MFCD00042653 | |
Myristic Acid Sodium Salt | |
sodium;tetradecanoate |
Safety and Handling
EINECSNumber : (2)-0611&(7)-0973
RTECSNumber : QH4455000
TSCA : Yes