missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Myristate 98.0+%, TCI America™

$103.10 - $869.43

Chemical Identifiers

CAS 822-12-8
Molecular Formula C14H27NaO2
Molecular Weight (g/mol) 250.358
MDL Number MFCD00042653
InChI Key JUQGWKYSEXPRGL-UHFFFAOYSA-M
Synonym Myristic Acid Sodium Salt
PubChem CID 23693095
IUPAC Name sodium;tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)[O-].[Na+]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M048325G
View Documents
TCI America
M048325G
25 g
Each for $103.10
 
M0483100G
View Documents
TCI America
M0483100G
100 g
Each for $276.63
 
M0483500G
View Documents
TCI America
M0483500G
500 g
Each for $869.43
 
Description

Description

Specifications

Chemical Identifiers

822-12-8
250.358
JUQGWKYSEXPRGL-UHFFFAOYSA-M
23693095
CCCCCCCCCCCCCC(=O)[O-].[Na+]
C14H27NaO2
MFCD00042653
Myristic Acid Sodium Salt
sodium;tetradecanoate

Specifications

White
Crystalline Powder
C14H27NaO2
Myristic Acid Sodium Salt
CCCCCCCCCCCCCC(=O)[O-].[Na+]
250.358
250.36
Sodium Myristate
822-12-8
25 g
MFCD00042653
JUQGWKYSEXPRGL-UHFFFAOYSA-M
sodium;tetradecanoate
23693095
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0611&(7)-0973

RTECSNumber : QH4455000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'