missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium oxalate, For ACS analysis, 99.96%, MP Biomedicals™
Supplier: MP Biomedicals Inc 0219143980
Specifications
Sodium oxalate | |
62-76-0 | |
2.34 (water = 1) | |
C2Na2O4 | |
100 g | |
ZNCPFRVNHGOPAG-UHFFFAOYSA-L | |
disodium oxalate | |
6125 | |
Crystalline Powder |
For ACS analysis | |
White | |
0.9996 | |
MFCD00012465 | |
sodium oxalate, disodium oxalate, natriumoxalat, ethanedioic acid, disodium salt, oxalic acid, disodium salt, natriumoxalat german, stavelan sodny czech, oxalic acid disodium salt, unii-7u0v68lt9x, ethanedioic acid disodium salt | |
[Na+].[Na+].[O-]C(=O)C([O-])=O | |
134 | |
134 |
Chemical Identifiers
62-76-0 | |
134 | |
ZNCPFRVNHGOPAG-UHFFFAOYSA-L | |
6125 | |
[Na+].[Na+].[O-]C(=O)C([O-])=O |
C2Na2O4 | |
MFCD00012465 | |
sodium oxalate, disodium oxalate, natriumoxalat, ethanedioic acid, disodium salt, oxalic acid, disodium salt, natriumoxalat german, stavelan sodny czech, oxalic acid disodium salt, unii-7u0v68lt9x, ethanedioic acid disodium salt | |
disodium oxalate |