missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium perborate tetrahydrate, 97%, pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 223200050

 View more versions of this product

Catalog No. AC223200050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Specifications

Sodium perborate tetrahydrate
White
10.1 to 10.6 (1% soln.)
5 kg
BNaO3·4H2O
01,1102; 02,387; 04,59; 14,290; 16,310
Solubility in water: 21.5g/L (18°C)
153.86
Pure
60.0°C
10486-00-7
Crystalline Powder
97%
NaBO3·4H2O
14,8652
153.86
97%
Plastic drum

Chemical Identifiers

10486-00-7
BNaO3·4H2O
153.86
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.
Harmful if inhaled.
May cause respiratory irritation.
May damage the unborn child.
Suspected of damaging fertility.

GHS P Statement
Do not handle until all safety precautions have been read and understood.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minu

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only