missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium perborate tetrahydrate, 97%, pure, Thermo Scientific Chemicals

$67.57 - $864.89

Chemical Identifiers

CAS 10486-00-7
Molecular Formula BNaO3·4H2O
Molecular Weight (g/mol) 153.86
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC223200250
SDS View Documents
Thermo Scientific Chemicals
223200250
25 g Plastic bottle
Each for $67.57
 
AC223202500
SDS View Documents
Thermo Scientific Chemicals
223202500
250 g Plastic bottle
Each for $95.61
 
AC223200010
SDS View Documents
Thermo Scientific Chemicals
223200010
1 kg Plastic bottle
Each for $226.41
 
AC223200050
SDS View Documents
Thermo Scientific Chemicals
223200050
5 kg Plastic drum
Each for $864.89
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Chemical Identifiers

10486-00-7
BNaO3·4H2O
153.86

Specifications

60.0°C
10486-00-7
Crystalline Powder
97%
NaBO3·4H2O
14,8652
153.86
97%
Plastic bottle
White
10.1 to 10.6 (1% soln.)
25 g
BNaO3·4H2O
01,1102; 02,387; 04,59; 14,290; 16,310
Solubility in water: 21.5g/L (18°C)
153.86
Pure
Sodium perborate tetrahydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.
Harmful if inhaled.
May cause respiratory irritation.
May damage the unborn child.
Suspected of damaging fertility.

GHS P Statement
Do not handle until all safety precautions have been read and understood.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minu

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only