Learn More
Sodium perborate tetrahydrate, 97%, pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 223200250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Specifications
Sodium perborate tetrahydrate | |
White | |
10.1 to 10.6 (1% soln.) | |
25 g | |
BNaO3·4H2O | |
01,1102; 02,387; 04,59; 14,290; 16,310 | |
Solubility in water: 21.5g/L (18°C) | |
153.86 | |
Pure |
60.0°C | |
10486-00-7 | |
Crystalline Powder | |
97% | |
NaBO3·4H2O | |
14,8652 | |
153.86 | |
97% | |
Plastic bottle |
Chemical Identifiers
10486-00-7 | |
BNaO3·4H2O | |
153.86 |
Safety and Handling
GHS H Statement
Causes serious eye damage.
Harmful if inhaled.
May cause respiratory irritation.
May damage the unborn child.
Suspected of damaging fertility.
GHS P Statement
Do not handle until all safety precautions have been read and understood.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minu
GHS Signal Word: Danger
RUO – Research Use Only