missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium perchlorate, ACS reagent, anhydrous, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 447421000

 View more versions of this product

Catalog No. AC447421000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium perchlorate
7601-89-0
ClNaO4
15,8784
Solubility in water: 209 g/100mL (15°C). Other solubilities: 170 g/100mL water (0°C), 300 g/100mL water (80°C), soluble in alcohol
[O-]Cl(=O)(=O)=O.[Na+]
122.44
122.44
Plastic bottle
482.0°C
100 g
NaClO4
sodium perchlorate, perchloric acid, sodium salt, natriumperchlorat, perchlorate de sodium, natriumperchloraat dutch, natriumperchlorat german, unii-97f4mty3va, cpl 46, perchlorate de sodium french, sodio perclorato di italian
BAZAXWOYCMUHIX-UHFFFAOYSA-M
sodium;perchlorate
522606
ACS Reagent

Chemical Identifiers

7601-89-0
122.44
sodium perchlorate, perchloric acid, sodium salt, natriumperchlorat, perchlorate de sodium, natriumperchloraat dutch, natriumperchlorat german, unii-97f4mty3va, cpl 46, perchlorate de sodium french, sodio perclorato di italian
sodium;perchlorate
ClNaO4
BAZAXWOYCMUHIX-UHFFFAOYSA-M
522606
[O-]Cl(=O)(=O)=O.[Na+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause fire or explosion; strong oxidizer.
Harmful if swallowed.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.

GHS Signal Word: Danger

EINECSNumber : 231-511-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only