missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium perchlorate, ACS reagent, anhydrous, Thermo Scientific Chemicals
$113.66 - $1044.27
Chemical Identifiers
CAS | 7601-89-0 |
---|---|
Molecular Formula | ClNaO4 |
Molecular Weight (g/mol) | 122.44 |
InChI Key | BAZAXWOYCMUHIX-UHFFFAOYSA-M |
Synonym | sodium perchlorate, perchloric acid, sodium salt, natriumperchlorat, perchlorate de sodium, natriumperchloraat dutch, natriumperchlorat german, unii-97f4mty3va, cpl 46, perchlorate de sodium french, sodio perclorato di italian |
PubChem CID | 522606 |
IUPAC Name | sodium;perchlorate |
SMILES | [O-]Cl(=O)(=O)=O.[Na+] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC447421000
|
Thermo Scientific Chemicals
447421000 |
100 g | Plastic bottle |
Each for $113.66
|
|
|||||
AC447425000
|
Thermo Scientific Chemicals
447425000 |
500 g | Plastic bottle |
Each for $326.81
|
|
|||||
AC447420025
|
Thermo Scientific Chemicals
447420025 |
2.5 kg | Plastic bottle |
Each for $1,044.27
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7601-89-0 | |
122.44 | |
sodium perchlorate, perchloric acid, sodium salt, natriumperchlorat, perchlorate de sodium, natriumperchloraat dutch, natriumperchlorat german, unii-97f4mty3va, cpl 46, perchlorate de sodium french, sodio perclorato di italian | |
sodium;perchlorate |
ClNaO4 | |
BAZAXWOYCMUHIX-UHFFFAOYSA-M | |
522606 | |
[O-]Cl(=O)(=O)=O.[Na+] |
Specifications
482.0°C | |
100 g | |
NaClO4 | |
sodium perchlorate, perchloric acid, sodium salt, natriumperchlorat, perchlorate de sodium, natriumperchloraat dutch, natriumperchlorat german, unii-97f4mty3va, cpl 46, perchlorate de sodium french, sodio perclorato di italian | |
BAZAXWOYCMUHIX-UHFFFAOYSA-M | |
sodium;perchlorate | |
522606 | |
ACS Reagent | |
Sodium perchlorate |
7601-89-0 | |
ClNaO4 | |
15,8784 | |
Solubility in water: 209 g/100mL (15°C). Other solubilities: 170 g/100mL water (0°C), 300 g/100mL water (80°C), soluble in alcohol | |
[O-]Cl(=O)(=O)=O.[Na+] | |
122.44 | |
122.44 | |
Plastic bottle |
Safety and Handling
GHS H Statement
May cause fire or explosion; strong oxidizer.
Harmful if swallowed.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
GHS Signal Word: Danger
EINECSNumber : 231-511-9
RUO – Research Use Only