missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Perchlorate Monohydrate (HPLC), Fisher Chemical™

$392.75 - $765.92

Chemical Identifiers

CAS 7791-07-3
Molecular Formula ClH2NaO5
Molecular Weight (g/mol) 140.45
MDL Number MFCD00149164
InChI Key IXGNPUSUVRTQGW-UHFFFAOYSA-M
Synonym sodium perchlorate monohydrate, sodium perchlorate hydrate, unii-g4h485lhcw, g4h485lhcw, ccris 8893, perchloric acid, sodium salt, monohydrate, sodium hydrate perchlorate, clo4.na.h2o, acmc-1bii1, sodium perchlorate-monohydrate
PubChem CID 516899
IUPAC Name sodium;perchlorate;hydrate
SMILES O.[O-]Cl(=O)(=O)=O.[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
S490-100
SDS View Documents Promotion Details
Thermo Fisher Scientific
S490100
100 g Glass Bottle
Each for $392.75
Case of 6 Each for $2,356.50
 
S490-500
SDS View Documents
Thermo Fisher Scientific
S490500
500 g Glass Bottle
Each for $765.92
Case of 6 Each for $4,595.52
 
Description

Description

Specifications

Chemical Identifiers

7791-07-3
140.45
IXGNPUSUVRTQGW-UHFFFAOYSA-M
516899
O.[O-]Cl(=O)(=O)=O.[Na+]
ClH2NaO5
MFCD00149164
sodium perchlorate monohydrate, sodium perchlorate hydrate, unii-g4h485lhcw, g4h485lhcw, ccris 8893, perchloric acid, sodium salt, monohydrate, sodium hydrate perchlorate, clo4.na.h2o, acmc-1bii1, sodium perchlorate-monohydrate
sodium;perchlorate;hydrate

Specifications

130°C
7791-07-3
Solid
ClH2NaO5
sodium perchlorate monohydrate, sodium perchlorate hydrate, unii-g4h485lhcw, g4h485lhcw, ccris 8893, perchloric acid, sodium salt, monohydrate, sodium hydrate perchlorate, clo4.na.h2o, acmc-1bii1, sodium perchlorate-monohydrate
O.[O-]Cl(=O)(=O)=O.[Na+]
140.45
140.46
Pass Test
2.02g/cm³
White
6.0 to 8.0
100 g
MFCD00149164
IXGNPUSUVRTQGW-UHFFFAOYSA-M
sodium;perchlorate;hydrate
516899
HPLC
Glass Bottle
Sodium Perchlorate Monohydrate
Videos
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 5.1, : Oxidizer

SDS
missing translation for 'documents'

missing translation for 'documents'