missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Perchlorate Monohydrate (HPLC), Fisher Chemical™

Supplier:  Thermo Fisher Scientific S490100

 View more versions of this product

Catalog No. S490-100



Explore available promotions

Description

Description

Specifications

Specifications

Sodium Perchlorate Monohydrate
White
6.0 to 8.0
100 g
MFCD00149164
IXGNPUSUVRTQGW-UHFFFAOYSA-M
sodium;perchlorate;hydrate
516899
HPLC
Glass Bottle
130°C
7791-07-3
Solid
ClH2NaO5
sodium perchlorate monohydrate, sodium perchlorate hydrate, unii-g4h485lhcw, g4h485lhcw, ccris 8893, perchloric acid, sodium salt, monohydrate, sodium hydrate perchlorate, clo4.na.h2o, acmc-1bii1, sodium perchlorate-monohydrate
O.[O-]Cl(=O)(=O)=O.[Na+]
140.45
140.46
Pass Test
2.02g/cm³

Chemical Identifiers

7791-07-3
140.45
IXGNPUSUVRTQGW-UHFFFAOYSA-M
516899
O.[O-]Cl(=O)(=O)=O.[Na+]
ClH2NaO5
MFCD00149164
sodium perchlorate monohydrate, sodium perchlorate hydrate, unii-g4h485lhcw, g4h485lhcw, ccris 8893, perchloric acid, sodium salt, monohydrate, sodium hydrate perchlorate, clo4.na.h2o, acmc-1bii1, sodium perchlorate-monohydrate
sodium;perchlorate;hydrate
Videos
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 5.1, : Oxidizer

SDS Documents

Documents

Promotions

Promotions