missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Phosphate Dibasic Anhydrous (Granular or Powder/Certified ACS), Fisher Chemical™
$368.67 - $11400.08
Chemical Identifiers
CAS | 7558-79-4 |
---|---|
Molecular Formula | HNa2O4P |
Molecular Weight (g/mol) | 141.96 |
MDL Number | MFCD00003496 |
InChI Key | BNIILDVGGAEEIG-UHFFFAOYSA-L |
Synonym | disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt |
PubChem CID | 24203 |
ChEBI | CHEBI:34683 |
IUPAC Name | disodium hydrogen phosphate |
SMILES | [Na+].[Na+].OP([O-])([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
S374-500
|
Thermo Fisher Scientific
S374500 |
500 g | Glass Bottle |
Each for $368.67
Case of 6 Each for $2,212.02
|
|
||||
S374-1
|
Thermo Fisher Scientific
S3741 |
1 kg | Poly Bottle |
Each for $469.25
Case of 6 Each for $2,815.50
|
|
||||
S374-3
|
Thermo Fisher Scientific
S3743 |
3 kg | Glass Bottle |
Each for $1,016.25
Case of 4 Each for $4,065.00
|
|
||||
S374-50
|
Thermo Fisher Scientific
S37450 |
50 kg | Fiber Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
7558-79-4 | |
141.96 | |
BNIILDVGGAEEIG-UHFFFAOYSA-L | |
24203 | |
disodium hydrogen phosphate |
HNa2O4P | |
MFCD00003496 | |
disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt | |
CHEBI:34683 | |
[Na+].[Na+].OP([O-])([O-])=O |
Specifications
7558-79-4 | |
8.7 to 9.3 | |
500 g | |
HNa2O4P | |
disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt | |
[Na+].[Na+].OP([O-])([O-])=O | |
141.96 | |
CHEBI:34683 | |
≥99% | |
Pass Test | |
Glass Bottle | |
Sodium Phosphate Dibasic Anhydrous |
White | |
Powder/Solid | |
≥99 % | |
MFCD00003496 | |
BNIILDVGGAEEIG-UHFFFAOYSA-L | |
disodium hydrogen phosphate | |
24203 | |
141.96 | |
Certified ACS | |
0.2% max. | |
Decomposes information: > 240°C |
Safety and Handling
CAUTION!
Emergency Overview
May cause eye, skin, and respiratory tract irritation . The toxicological properties have not been fully investigated. Hygroscopic.
NFPA
Health:1 Flammability:0 Instability:1
Signal Word: CAUTION!