missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Phosphate Dibasic Anhydrous (Granular or Powder/Certified ACS), Fisher Chemical™

Supplier:  Fisher Chemical S3743

 View more versions of this product

Catalog No. S374-3



Explore available promotions

Description

Description

Specifications

Specifications

Sodium Phosphate Dibasic Anhydrous
White
Powder/Solid
≥99 %
MFCD00003496
BNIILDVGGAEEIG-UHFFFAOYSA-L
disodium hydrogen phosphate
24203
141.96
Certified ACS
0.2% max.
Decomposes information: > 240°C
7558-79-4
8.7 to 9.3
3 kg
HNa2O4P
disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
[Na+].[Na+].OP([O-])([O-])=O
141.96
CHEBI:34683
≥99%
Pass Test
Glass Bottle

Chemical Identifiers

7558-79-4
141.96
BNIILDVGGAEEIG-UHFFFAOYSA-L
24203
disodium hydrogen phosphate
HNa2O4P
MFCD00003496
disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
CHEBI:34683
[Na+].[Na+].OP([O-])([O-])=O
Videos
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause eye, skin, and respiratory tract irritation . The toxicological properties have not been fully investigated. Hygroscopic.
NFPA
Health:1 Flammability:0 Instability:1

Signal Word: CAUTION!

SDS Documents

Documents

Promotions

Promotions