missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Phosphate Monobasic Monohydrate (Crystalline/Certified ACS), Fisher Chemical™

Supplier:  Fisher Chemical S3693

 View more versions of this product

Catalog No. S369-3



Explore available promotions

Description

Description

Specifications

Specifications

Sodium Phosphate Monobasic Monohydrate
White
4.1 to 4.5
3 kg
98.0-102.0%
MFCD00149208
BBMHARZCALWXSL-UHFFFAOYSA-N
sodium phosphoric acid hydrate
516949
98 to 102%
Pass Test
100°C
10049-21-5
Solid
98 to 102 %
H5NaO5P
sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
O.[Na+].OP(O)(O)=O
139.00
137.99
Certified ACS
Poly Bottle

Chemical Identifiers

10049-21-5
139.00
BBMHARZCALWXSL-UHFFFAOYSA-N
516949
O.[Na+].OP(O)(O)=O
H5NaO5P
MFCD00149208
sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
sodium phosphoric acid hydrate
Safety and Handling

Safety and Handling

SDS Documents

Documents

Promotions

Promotions