missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Monobasic Monohydrate (Crystalline/Certified ACS), Fisher Chemical™

$398.92 - $8696.83

Chemical Identifiers

CAS 10049-21-5
Molecular Formula H5NaO5P
Molecular Weight (g/mol) 139.00
MDL Number MFCD00149208
InChI Key BBMHARZCALWXSL-UHFFFAOYSA-N
Synonym sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
PubChem CID 516949
IUPAC Name sodium phosphoric acid hydrate
SMILES O.[Na+].OP(O)(O)=O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
S369-500
SDS View Documents Promotion Details
Fisher Chemical
S369500
500 g Poly Bottle
Each for $398.92
Case of 6 Each for $2,393.52
Only null left
 
S369-1
SDS View Documents Promotion Details
Thermo Fisher Scientific
S3691
1 kg Poly Bottle
Each for $434.67
Case of 6 Each for $2,608.02
Only null left
 
S369-3
SDS View Documents Promotion Details
Fisher Chemical
S3693
3 kg Poly Bottle
Each for $1,335.50
Case of 4 Each for $5,342.00
Only null left
 
S369-50
SDS Accessories View Documents Promotion Details
Fisher Chemical
S36950
50 kg Fiber Drum
Each for $8,696.83
Only null left
 
Description

Description

Specifications

Chemical Identifiers

10049-21-5
139.00
BBMHARZCALWXSL-UHFFFAOYSA-N
516949
O.[Na+].OP(O)(O)=O
H5NaO5P
MFCD00149208
sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
sodium phosphoric acid hydrate

Specifications

100°C
10049-21-5
Solid
98 to 102 %
MFCD00149208
BBMHARZCALWXSL-UHFFFAOYSA-N
sodium phosphoric acid hydrate
516949
98 to 102%
Pass Test
Sodium Phosphate Monobasic Monohydrate
White
4.1 to 4.5
500 g
H5NaO5P
sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
O.[Na+].OP(O)(O)=O
139.00
137.99
Certified ACS
Poly Bottle
98.0-102.0%
Videos
Safety and Handling

Safety and Handling

SDS
missing translation for 'documents'

missing translation for 'documents'