missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Phosphate Monobasic Monohydrate (Crystalline/Certified ACS), Fisher Chemical™
Supplier: Thermo Fisher Scientific S36950
Specifications
Sodium Phosphate Monobasic Monohydrate | |
White | |
4.1 to 4.5 | |
50 kg | |
98.0-102.0% | |
MFCD00149208 | |
BBMHARZCALWXSL-UHFFFAOYSA-N | |
sodium phosphoric acid hydrate | |
516949 | |
98 to 102% | |
Pass Test |
100°C | |
10049-21-5 | |
Solid | |
98 to 102 % | |
H5NaO5P | |
sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707 | |
O.[Na+].OP(O)(O)=O | |
139.00 | |
137.99 | |
Certified ACS | |
Fiber Drum |
Chemical Identifiers
10049-21-5 | |
139.00 | |
BBMHARZCALWXSL-UHFFFAOYSA-N | |
516949 | |
O.[Na+].OP(O)(O)=O |
H5NaO5P | |
MFCD00149208 | |
sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707 | |
sodium phosphoric acid hydrate |