missing translation for 'onlineSavingsMsg'
Learn More

Sodium polyphosphate, pure

CAS: 50813-16-6 | (NaO3P)n

$83.43 - $437.59

Chemical Identifiers

CAS 50813-16-6
Molecular Formula (NaO3P)n
MDL Number MFCD00134118
Synonym hexasodium oxido oxido phosphonatooxy phosphoryl oxy phosphoryl oxy phospho phosphonato oxy phosphinate
IUPAC Name Sodium polyphosphate
SMILES [Na]OP(-*)(=O)O-*
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC390932500
SDS View Documents
Thermo Scientific Chemicals
390932500
250 g Plastic bottle N/A
Only null left
 
AC390930010
SDS View Documents
Thermo Scientific Chemicals
390930010
1 kg Plastic bottle N/A
Only null left
 
AC390930050
SDS View Documents
Thermo Scientific Chemicals
390930050
5 kg Plastic bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

50813-16-6
MFCD00134118
Sodium polyphosphate
(NaO3P)n
hexasodium oxido oxido phosphonatooxy phosphoryl oxy phosphoryl oxy phospho phosphonato oxy phosphinate
[Na]OP(-*)(=O)O-*

Specifications

600.0°C
50813-16-6
250 g
(NaPO3)n
15, 8773
Solubility in water: 1000g/l (20°C)
Sodium polyphosphate
Pure
Sodium polyphosphate
White
Crystalline Powder
(NaO3P)n
MFCD00134118
hexasodium oxido oxido phosphonatooxy phosphoryl oxy phosphoryl oxy phospho phosphonato oxy phosphinate
[Na]OP(-*)(=O)O-*
65 to 70%
Plastic bottle
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 256-779-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only