missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Propionate, NF, 99-100.5%, Spectrum™ Chemical

S1418, 6700-17-0, C3H5NaO2·xH2O

Supplier:  Spectrum Chemical Mfg Cor S1418500GM

 View more versions of this product

Catalog No. 18606760


Explore available promotions

Description

Description

Spectrum™ Chemical Sodium Propionate, NF is used as an antiseptic agent. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Specifications

6700-17-0
Poly Pail
500 g
O.[Na+].CCC([O-])=O
114.08
NF
1
C3H7NaO3
HOAUAOBUGFYWMK-UHFFFAOYSA-M
sodium hydrate propanoate
99 to 100.5%

Chemical Identifiers

6700-17-0
114.08
sodium hydrate propanoate
C3H7NaO3
HOAUAOBUGFYWMK-UHFFFAOYSA-M
O.[Na+].CCC([O-])=O
Videos
SDS
Documents

Documents

Promotions

Promotions