missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Propionate, NF, 99-100.5%, Spectrum™ Chemical
S1418, 6700-17-0, C3H5NaO2·xH2O
$386.66 - $9,718.49
Chemical Identifiers
CAS | 6700-17-0 |
---|---|
Molecular Formula | C3H7NaO3 |
Molecular Weight (g/mol) | 114.08 |
InChI Key | HOAUAOBUGFYWMK-UHFFFAOYSA-M |
IUPAC Name | sodium hydrate propanoate |
SMILES | O.[Na+].CCC([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18606760
|
Spectrum Chemical Mfg Cor
S1418500GM |
500 g | Poly Pail |
N/A
|
|
||||
18606761
|
Spectrum Chemical Mfg Cor
S141825KG |
2.5 kg | Poly Pail |
N/A
|
|
||||
18606762
|
Spectrum Chemical Mfg Cor
S141812KG |
12 kg | Fiber Drum |
N/A
|
|
||||
18606763
|
Spectrum Chemical Mfg Cor
S141845KGBL |
45 kg | Fiber Drum |
N/A
|
|
||||
Description
Spectrum™ Chemical Sodium Propionate, NF is used as an antiseptic agent. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
6700-17-0 | |
114.08 | |
sodium hydrate propanoate |
C3H7NaO3 | |
HOAUAOBUGFYWMK-UHFFFAOYSA-M | |
O.[Na+].CCC([O-])=O |
Specifications
6700-17-0 | |
Poly Pail | |
500 g | |
O.[Na+].CCC([O-])=O | |
114.08 | |
NF |
1 | |
C3H7NaO3 | |
HOAUAOBUGFYWMK-UHFFFAOYSA-M | |
sodium hydrate propanoate | |
99 to 100.5% |