missing translation for 'onlineSavingsMsg'
Learn More

Sodium selenate, 98%

CAS: 13410-01-0 | Na2O4Se | 188.95 g/mol

$200.60 - $1,432.99

Chemical Identifiers

CAS 13410-01-0
Molecular Formula Na2O4Se
Molecular Weight (g/mol) 188.95
MDL Number MFCD00003490
InChI Key MHQOTKLEMKRJIR-UHFFFAOYSA-L
Synonym sodium selenate, disodium selenate, natriumseleniat, selenic acid, disodium salt, caswell no. 791, natriumseleniat german, sel-tox sso2 and ss-20, unii-5dqp25600a, selenic acid h2seo4 , disodium salt, ccris 1259
PubChem CID 25960
ChEBI CHEBI:77775
IUPAC Name disodium;selenate
SMILES [Na+].[Na+].[O-][Se]([O-])(=O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC369960250
SDS View Documents
Thermo Scientific Chemicals
369960250
25 g Glass bottle N/A
Only null left
 
AC369961000
SDS View Documents
Thermo Scientific Chemicals
369961000
100 g Glass bottle N/A
Only null left
 
AC369965000
SDS View Documents
Thermo Scientific Chemicals
369965000
500 g Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13410-01-0
188.95
MHQOTKLEMKRJIR-UHFFFAOYSA-L
25960
disodium;selenate
Na2O4Se
MFCD00003490
sodium selenate, disodium selenate, natriumseleniat, selenic acid, disodium salt, caswell no. 791, natriumseleniat german, sel-tox sso2 and ss-20, unii-5dqp25600a, selenic acid h2seo4 , disodium salt, ccris 1259
CHEBI:77775
[Na+].[Na+].[O-][Se]([O-])(=O)=O

Specifications

13410-01-0
Crystalline Powder
97.5% min
MFCD00003490
sodium selenate, disodium selenate, natriumseleniat, selenic acid, disodium salt, caswell no. 791, natriumseleniat german, sel-tox sso2 and ss-20, unii-5dqp25600a, selenic acid h2seo4 , disodium salt, ccris 1259
MHQOTKLEMKRJIR-UHFFFAOYSA-L
disodium;selenate
25960
188.94
confirmed
Sodium selenate
White
25 g
Na2O4Se
15, 8800
Solubility in water: 19g/L (20°C)
[Na+].[Na+].[O-][Se]([O-])(=O)=O
188.95
CHEBI:77775
98%
Glass bottle
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Fatal if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED:

GHS Signal Word: Danger

EINECSNumber : 236-501-8

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only