missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium sulfate, 99+%, ACS reagent, granular, anhydrous, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 354250010

 View more versions of this product

Catalog No. AC354250010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

>1700.0°C
884.0°C
7757-82-6
5.2 to 7.0 (5% soln. at 25°C)
1 kg
Na2O4S
MFCD00003504
15, 8809
PMZURENOXWZQFD-UHFFFAOYSA-L
disodium sulfate
24436
142.04
ACS Reagent
0.5 % max.
Decomposition Information: >890°C
Sodium sulfate
Anhydrous
Colorless to Orange
Granules
99+%
Na2SO4
01,1103
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
[Na+].[Na+].[O-]S([O-])(=O)=O
142.04
CHEBI:32149
≥99%
Sodium : passes test (USP) Sulfate : passes test (USP)
Plastic bottle
2.68g/mL

Chemical Identifiers

7757-82-6
142.04
PMZURENOXWZQFD-UHFFFAOYSA-L
24436
[Na+].[Na+].[O-]S([O-])(=O)=O
Na2O4S
MFCD00003504
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
CHEBI:32149
Videos
Safety and Handling

Safety and Handling

Emergency Overview
May cause skin, eye, and respiratory tract irritation. The toxicological properties have not been fully investigated. Hygroscopic.
NFPA
Health: 1
Flammability: 1
Instability: 0

SIGNAL WORD: CAUTION!

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only