missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium sulfate, 99+%, ACS reagent, granular, anhydrous, Thermo Scientific Chemicals
$173.42 - $641.28
Chemical Identifiers
CAS | 7757-82-6 |
---|---|
Molecular Formula | Na2O4S |
Molecular Weight (g/mol) | 142.04 |
MDL Number | MFCD00003504 |
InChI Key | PMZURENOXWZQFD-UHFFFAOYSA-L |
Synonym | sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1 |
PubChem CID | 24436 |
ChEBI | CHEBI:32149 |
SMILES | [Na+].[Na+].[O-]S([O-])(=O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC354250010
|
Thermo Scientific Chemicals
354250010 |
1 kg | Plastic bottle |
N/A
|
|
||||
AC354250025
|
Thermo Scientific Chemicals
354250025 |
2.5 kg | Plastic bottle |
N/A
|
|
||||
AC354250100
|
Thermo Scientific Chemicals
354250100 |
10 kg | Plastic drum |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7757-82-6 | |
142.04 | |
PMZURENOXWZQFD-UHFFFAOYSA-L | |
24436 | |
[Na+].[Na+].[O-]S([O-])(=O)=O |
Na2O4S | |
MFCD00003504 | |
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1 | |
CHEBI:32149 |
Specifications
>1700.0°C | |
7757-82-6 | |
5.2 to 7.0 (5% soln. at 25°C) | |
1 kg | |
Na2O4S | |
MFCD00003504 | |
15, 8809 | |
PMZURENOXWZQFD-UHFFFAOYSA-L | |
disodium sulfate | |
24436 | |
142.04 | |
ACS Reagent | |
0.5 % max. | |
Decomposition Information: >890°C | |
Sodium sulfate, Anhydrous |
884.0°C | |
Colorless to Orange | |
Granules | |
99+% | |
Na2SO4 | |
01,1103 | |
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1 | |
[Na+].[Na+].[O-]S([O-])(=O)=O | |
142.04 | |
CHEBI:32149 | |
≥99% | |
Sodium : passes test (USP) Sulfate : passes test (USP) | |
Plastic bottle | |
2.68g/mL |
Safety and Handling
Emergency Overview
May cause skin, eye, and respiratory tract irritation. The toxicological properties have not been fully investigated. Hygroscopic.
NFPA
Health: 1
Flammability: 1
Instability: 0
SIGNAL WORD: CAUTION!
EINECSNumber : 231-820-9
RUO – Research Use Only