missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing

Sodium Sulfate Anhydrous (Granular/Certified ACS), Fisher Chemical™
$246.00 - $17108.33
Chemical Identifiers
CAS | 7757-82-6 |
---|---|
Molecular Formula | Na2O4S |
Molecular Weight (g/mol) | 142.04 |
MDL Number | MFCD00003504 |
InChI Key | PMZURENOXWZQFD-UHFFFAOYSA-L |
Synonym | sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1 |
PubChem CID | 24436 |
ChEBI | CHEBI:32149 |
IUPAC Name | disodium sulfate |
SMILES | [Na+].[Na+].[O-]S([O-])(=O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
S421-500
![]() |
Thermo Fisher Scientific
S421500 |
500 g | Glass Bottle |
Each for $246.00
Case of 6 Each for $1,476.00
|
![]() |
|||||
S421-300LB
|
Thermo Fisher Scientific
FLS421300LB |
300 lb. | Fiber Drum |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | ||||||||||
S421-1
![]() |
Thermo Fisher Scientific
S4211 |
1 kg | Poly Bottle |
Each for $336.92
Case of 6 Each for $2,021.52
|
![]() |
|||||
S421-3
|
Thermo Fisher Scientific
S4213 |
3 kg | Glass Bottle |
Each for $756.33
Case of 4 Each for $3,025.32
|
|
|||||
S421-10
|
Thermo Fisher Scientific
S42110 |
10 kg | Poly Pail |
Each for $1,829.92
|
|
|||||
S421-50
|
Thermo Fisher Scientific
S42150 |
50 kg | Fiber Drum |
Each for $7,895.17
|
|
|||||
Chemical Identifiers
7757-82-6 | |
142.04 | |
PMZURENOXWZQFD-UHFFFAOYSA-L | |
24436 | |
disodium sulfate |
Na2O4S | |
MFCD00003504 | |
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1 | |
CHEBI:32149 | |
[Na+].[Na+].[O-]S([O-])(=O)=O |
Specifications
884°C | |
White | |
Solid | |
≥99.0 % | |
MFCD00003504 | |
PMZURENOXWZQFD-UHFFFAOYSA-L | |
disodium sulfate | |
24436 | |
142.04 | |
Certified ACS | |
0.5% max. | |
Decomposition Information: >890°C | |
≥99.0% |
7757-82-6 | |
5 to 8 | |
500 g | |
Na2O4S | |
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1 | |
[Na+].[Na+].[O-]S([O-])(=O)=O | |
142.04 | |
CHEBI:32149 | |
≥99.0% | |
Pass Test | |
Glass Bottle | |
Sodium Sulfate Anhydrous |
Safety and Handling

CAUTION!
Emergency Overview
May cause skin, eye, and respiratory tract irritation. The toxicological properties have not been fully investigated. Hygroscopic.
NFPA
Health: 1
Flammability: 1
Instability: 0
SIGNAL WORD: CAUTION!