missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
![](http://beta-static.fishersci.ca/images/Gray.gif)
Sodium Sulfate Anhydrous (Granular/Certified ACS), Fisher Chemical™
Supplier: Fisher Chemical S42110
Specifications
Sodium Sulfate Anhydrous | |
7757-82-6 | |
5 to 8 | |
10 kg | |
≥99.0% | |
MFCD00003504 | |
PMZURENOXWZQFD-UHFFFAOYSA-L | |
disodium sulfate | |
24436 | |
142.04 | |
Certified ACS | |
0.5% max. | |
Decomposition Information: >890°C |
884°C | |
White | |
Solid | |
≥99.0 % | |
Na2O4S | |
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1 | |
[Na+].[Na+].[O-]S([O-])(=O)=O | |
142.04 | |
CHEBI:32149 | |
≥99.0% | |
Pass Test | |
Poly Pail |
Chemical Identifiers
7757-82-6 | |
142.04 | |
PMZURENOXWZQFD-UHFFFAOYSA-L | |
24436 | |
disodium sulfate |
Na2O4S | |
MFCD00003504 | |
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1 | |
CHEBI:32149 | |
[Na+].[Na+].[O-]S([O-])(=O)=O |
Safety and Handling
![](http://beta-static.fishersci.ca/images/Gray.gif)
CAUTION!
Emergency Overview
May cause skin, eye, and respiratory tract irritation. The toxicological properties have not been fully investigated. Hygroscopic.
NFPA
Health: 1
Flammability: 1
Instability: 0
SIGNAL WORD: CAUTION!