missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium thiocyanate, ACS, 98.0% min, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 03338836

 View more versions of this product

Catalog No. AA3338836

Explore available promotions

Description

Description

Sodium thiocyanate is used as a precursor for the synthesis of pharmaceuticals and specialty chemicals. It is used to prepare alkylthiocyanates by reacting with alkyl halides. It is involved nucleophilic substitution reactions with phase-transfer catalysts. It is a common laboratory reagent used to test presence of Fe3+ ions. It acts as a spinning solvent for polyacrylonitrile in fiber industry, photographic stabilizer, corrosion inhibitor, polymer stabilizer and hardening accelerator for cement and concrete mixes. Further, it is also used in both the crystallization and purification of proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium thiocyanate
For analysis ACS
White
500 g
CNNaS
3594965
Soluble in water,acetone,alcohol.Soluble in water,acetone,alcohols and ammonia.
[Na+].[S-]C#N
81.07
CHEBI:30952
ACS
White crystalline/chunks
287°C
540-72-7
Crystals and/or Chunks
98% min.
MFCD00011123
sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased
VGTPCRGMBIAPIM-UHFFFAOYSA-M
sodium;thiocyanate
516871
81.07
Hygroscopic; Light sensitive
1.74 g/cm3 at 20°C

Chemical Identifiers

540-72-7
81.07
VGTPCRGMBIAPIM-UHFFFAOYSA-M
516871
[Na+].[S-]C#N
CNNaS
MFCD00011123
sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased
CHEBI:30952
Safety and Handling

Safety and Handling

P260-P264b-P270-P271-P280-P302+P352-P304+P340-P314-P330-P362-P501c

H302+H312+H332-H373

EINECSNumber : 208-754-4

RTECSNumber : XL2275000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only