missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Soft Sided Table Top Heating Mantle

Fisherbrand™ Soft Sided Table Top Heating Mantle is a self-supporting table top heating mantle, used to provide safe and uniform heat distribution for round-bottom flasks.

 View more versions of this product

Catalog No. 11-476-037


Explore available promotions

Requires : Temperature Controller

Description

Description

  • Fiberglass cloth out shell molded to fit round-bottom flasks
  • Max temperature up to 482°C (900°F)
  • Uniform heat distribution
  • Multi-stranded resistance wire heating element for increased durability and flexibility
  • Temperature controller required for use

Warranty

1-year

Specifications

Specifications

RoHs, REACH
Round Bottom Flasks, Temperature Controller
72 in. (1.8 m) Power Cord with Standard 2-Prong Plug (NEMA 1-15)
210 W
Max 900°F
500 mL
120 V
Fiberglass
482°C (Max.)
Round bottom flask heating
Videos
SDS
Documents

Documents

Promotions

Promotions