Sonoco™ ThermoSafe DiagnosticShipper Systems, Without Tubes
Ready-to-use combination/triple packaging kits
Supplier: Sonoco™ 474
Description
Tested and certified to meet the material and performance requirements for the transport of diagnostic specimens in conformance with U.S. Department of Transportation (DOT), U.S. Postal Service and International Air Transport Association (IATA) Packing Instruction 650.
Specifications
Absorbent material, tape, poly bag, carton | |
Conforms with DOT, USPS, IATA packing instruction 650 | |
Diagnostic specimen shipping system without tubes |