Sonoco™ ThermoSafe Envoy™ Global Pre-Qualified Insulated Shippers with Winter Packout
Temperature controlled packaging solution
$2458.88 - $2458.88
Catalog Number | Mfr. No. | Capacity (Metric) | Dimensions (L x W x H) Exterior | Temperature Range (Metric) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Dimensions (L x W x H) Exterior | Temperature Range (Metric) | Price | Quantity | |||
03-531-713
|
Sonoco™
E18C2W |
18.2 L | 521 x 483 x 527 mm (20.5 x 19 x 20.7 in.) | CRT: 15° to 25°C |
Case of 8 Each for $2,458.88
|
|
|||
Description
- Winter packout configuration
- Positioned as low material packaging cost solution
- Offers various sizes, temperature ranges and flexibility
- Envoy Global line is able to change from controlled room temperature, refrigerated and frozen temperatures by changing the refrigerants
- Simple and error-free
- Utilizes four EPS container sizes
- Pre-Qualified to ISCsilver ambient profile
- Zero Bench Time refrigerant design
- Expanded polystyrene insulation
- Low packaging material cost
- Simple designs
- Air flow channels built into EPS, allows for less refrigerant needed in each packout
Duration
- Refrigerated and CRT: Minimum of two days
- Frozen: Minimum of four days
Shipping pharmaceuticals, clinical treatments and diagnostic samples
Specifications
Expanded Polystyrene |