Fisherbrand™ Specific Gravity/Baume Hydrometers
Fisherbrand™ Specific Gravity/Baume Hydrometers are calibrated according to ASTM™ E 126 specifications.
 View more versions of this product
Catalog No. 11-532C
Explore available promotions
Description
Description
Fisherbrand™ Specific Gravity/Baume Hydrometers are calibrated according to ASTM™ E 126 specifications.
  • Permanently marked with individual serial numbers
Order Info
A certificate of calibration is available; simply add the letter C to the end of the catalog number when ordering.
Certifications
Certificate of Accuracy and Compliance provided to indicate calibration against standards provided by NIST™.
Compliance
Accuracy meets or exceeds ASTM™ E 100 specifications
Specifications
Specifications
Specific Gravity Hydrometer
NIST, ASTM E100, ASTM E126
175mL Borosilicate Glass cylinder
Measures liquids lighter than Water
140 mL
0.002
170 mm
Soda Lime Glass
0.900 to 1.000
Videos
SDS
Documents
Documents
Accessories