Fisherbrand™ Specific Gravity/Baume Hydrometers
Fisherbrand™ Specific Gravity/Baume Hydrometers are calibrated according to ASTM™ E 126 specifications.
$44.48 - $741.52
Catalog Number | Mfr. No. | Certifications/Compliance | For Use With (Equipment) | Specific Gravity Range | Volume (Metric) | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Certifications/Compliance | For Use With (Equipment) | Specific Gravity Range | Volume (Metric) | Price | Quantity | ||
11-540B
|
N/A | NIST, ASTM E100, ASTM E126 | 300mL Borosilicate Glass cylinder | 1.200 to 1.420 | 250 mL |
Each for $48.41
|
|
||
11-542D
|
N/A | NIST, ASTM E100, ASTM E126 | 300mL Borosilicate Glass cylinder | 1.000 to 2.000 | 140 mL |
Each for $44.48
|
|
||
11-542CC
|
N/A | 3-point Certification, NIST, ASTM E100, ASTM E126 | 300mL Borosilicate Glass cylinder | 1.400 to 1.600 | 140 mL |
Each for $653.55
|
|
||
11-540BC
|
N/A | 3-point Certification, NIST, ASTM E100, ASTM E126 | 300mL Borosilicate Glass cylinder | 1.200 to 1.420 | 250 mL |
Each for $741.52
|
|
||
11-540E
|
N/A | NIST, ASTM E100, ASTM E126 | 300mL Borosilicate Glass cylinder | 1.000 to 2.000 | 250 mL |
Each for $44.48
|
|
||
11-540EC
|
N/A | 3-point Certification, NIST, ASTM E100, ASTM E126 | 300mL Borosilicate Glass cylinder | 1.000 to 2.000 | 250 mL |
Each for $643.86
|
|
||
11-540A
|
N/A | NIST, ASTM E100, ASTM E126 | 300mL Borosilicate Glass cylinder | 1.000 to 1.220 | 250 mL |
Each for $47.76
|
|
||
11-540D
|
N/A | NIST, ASTM E100, ASTM E126 | 300mL Borosilicate Glass cylinder | 1.600 to 1.820 | 250 mL |
Each for $47.76
|
|
||
11-550
|
N/A | NIST, ASTM E100, ASTM E126 | 600mL Borosilicate Glass cylinder | 0.7 to 2 | 515 mL |
Case for $44.72
|
|
||
11-540AC
|
N/A | 3-point Certification, NIST, ASTM E100, ASTM E126 | 300mL Borosilicate Glass cylinder | 1.000 to 1.220 | 250 mL |
Each for $736.86
|
|
||
11-542C
|
N/A | NIST, ASTM E100, ASTM E126 | 300mL Borosilicate Glass cylinder | 1.400 to 1.600 | 140 mL |
Each for $47.76
|
|
||
11540CC
|
N/A | 3-point Certification, NIST, ASTM E100, ASTM E126 | 300mL Borosilicate Glass cylinder | 1.400 to 1.620 | 250 mL |
Each for $689.38
|
|
||
11-532B
|
N/A | NIST, ASTM E100, ASTM E126 | 175mL Borosilicate Glass cylinder | 0.800 to 0.900 | 140 mL |
Each for $48.66
|
|
||
11-532BC
|
N/A | 3-point Certification, NIST, ASTM E100, ASTM E126 | 175mL Borosilicate Glass cylinder | 0.800 to 0.900 | 140 mL |
Each for $735.41
|
|
||
11-542B
|
N/A | NIST, ASTM E100, ASTM E126 | 300mL Borosilicate Glass cylinder | 1.200 to 1.400 | 140 mL |
Each for $47.76
|
|
||
11-550C
|
N/A | 3-point Certification, NIST, ASTM E100, ASTM E126 | 600mL Borosilicate Glass cylinder | 0.700 to 2.000 | 515 mL |
Each for $673.28
|
|
||
11-542DC
|
N/A | 3-point Certification, NIST, ASTM E100, ASTM E126 | 300mL Borosilicate Glass cylinder | 1 to 2 | 140 mL |
Each for $671.86
|
|
||
11-532CC
|
N/A | 3-point Certification, NIST, ASTM E100, ASTM E126 | 175mL Borosilicate Glass cylinder | 0.900 to 1.000 | 140 mL |
Each for $656.17
|
|
||
11-532C
|
N/A | NIST, ASTM E100, ASTM E126 | 175mL Borosilicate Glass cylinder | 0.900 to 1.000 | 140 mL |
Each for $48.66
|
|
||
11-540C
|
N/A | NIST, ASTM E100, ASTM E126 | 300mL Borosilicate Glass cylinder | 1.400 to 1.620 | 250 mL |
Each for $47.76
|
|
||
11-542AC
|
N/A | 3-point Certification, NIST, ASTM E100, ASTM E126 | 300mL Borosilicate Glass cylinder | 1.000 to 1.225 | 140 mL |
Each for $661.59
|
|
||
11-542A
|
N/A | NIST, ASTM E100, ASTM E126 | 300mL Borosilicate Glass cylinder | 1.000 to 1.225 | 140 mL |
Each for $47.76
|
|
||
Description
Fisherbrand™ Specific Gravity/Baume Hydrometers are calibrated according to ASTM™ E 126 specifications.
- Permanently marked with individual serial numbers
Order Info
A certificate of calibration is available; simply add the letter C to the end of the catalog number when ordering.
Certifications
Certificate of Accuracy and Compliance provided to indicate calibration against standards provided by NIST™.
Compliance
Accuracy meets or exceeds ASTM™ E 100 specifications