FyterTech Nonwovens Spilfyter™ Chemical Classifiers Strip Kits
For identifying risks in unknown spilled liquids
Supplier:  FyterTech Nonwovens 571020
 View more versions of this product
Catalog No. 19-065-569
Explore available promotions
Description
Description
Chemical Classifier Strips show responder if an unknown spill contains critical levels of these contaminant categories: acid/base, oxidizer, fluoride, petroleum/organic solvent, iodine/bromine/chlorine. Classifier Chart compares sample colors to standard color.
  • Recommended for all emergency response teams
  • Quickly assist responder in classifying spill for proper treatment
  • Use anywhere
  • Case or tube versions
Chemical Classifier Kit consists of:
  • Six Chemical Classifier Strips
  • One bobbin of polypropylene thread
  • Laminated Chemical Classifier Chart
  • Carrying Tube
Chemical Classifier Station consists of:
  • 20 Chemical Classifier Strips
  • Two bobbins of polypropylene thread
  • Laminated Chemical Classifier Chart
  • Carrying Case
Use strips to identify unknowns in sewers, construction sites, accident sites, storage facilities, landfills, waste management facilities.
Order Info
Additional test strips and classifier charts are available separately.
Specifications
Specifications
Chemical Classifier Station
Chemical classifier strips, bobbin of polypropylene thread, laminated chemical classifier chart
20°C
1 kg
Identify unknowns in sewers, construction sites, accident sites, storage facilities, landfills, waste management facilities by using
1 Ea.
3 lb.