FyterTech Nonwovens Spilfyter™ KolorSafe™ Neutralizers for Acids
Take the guesswork out of neutralizing hazardous spills
Supplier:  FyterTech Nonwovens 410020
 View more versions of this product
Catalog No. 19-066-518
Explore available promotions
Description
Description
Unlike dry neutralizer, liquid reagent can be sprayed into hard-to-reach areas and on vertical surfaces.
  • Sodium-free liquid reagent changes color when neutralization is complete
  • Safe for use in laboratories and cleanrooms
Specifications
Specifications
5 gal.
Liquid Acid Neutralizer
Spill control
1 Ea.
22 kg
18.92 L
Liquid
Color change indicator when neutralized
48 lb.