missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
FyterTech Nonwovens Spilfyter™ KolorSafe™ Neutralizers for Acids
Take the guesswork out of neutralizing hazardous spills
$290.68 - $727.41
Specifications
For Use With (Application) | Spill control |
---|---|
Includes | Color change indicator when neutralized |
Catalog Number | Mfr. No. | Capacity (English) | Capacity (Metric) | Type | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (English) | Capacity (Metric) | Type | Price | Quantity | |||
19-066-518
|
FyterTech Nonwovens
410020 |
5 gal. | 18.92 L | Liquid Acid Neutralizer |
Each for $727.41
|
|
|||
19-021889
|
FyterTech Nonwovens
JV410004 |
1 gal. | 3.78 L | Acid Neutralizer |
Case of 4 Each for $322.61
|
|
|||
19-021888
|
FyterTech Nonwovens
JV410001 |
0.24 gal. | 0.94 L | Acid Neutralizer |
Case of 12 Each for $360.54
|
|
|||
19-021893
|
FyterTech Nonwovens
JV440001 |
2 lb. | 0.9 kg | Dry Acid Neutralizer |
Case of 10 Each for $290.68
|
|
|||
Description
Unlike dry neutralizer, liquid reagent can be sprayed into hard-to-reach areas and on vertical surfaces.
- Sodium-free liquid reagent changes color when neutralization is complete
- Safe for use in laboratories and cleanrooms
Specifications
Spill control | |
Color change indicator when neutralized |