FyterTech Nonwovens Spilfyter™ Wastewater Classifiers
Distinguish risks of unknown spilled liquids
Supplier:  FyterTech Nonwovens 580001
 View more versions of this product
Catalog No. 19-120-875
Explore available promotions
Description
Description
Classifier Strips show responder if unknown spill contains critical levels of these contaminant categories: acid/base; petroleum/organic solvent; hydrogen sulfide; nitrite; nitrate; fluoride. Classifier chart compares standard colors to sample colors.
Wastewater Classifier Kit consists of:
  • Six Wastewater Classifier Strips
  • One bobbin of polypropylene thread
  • Laminated Chemical Classifier Chart
Wastewater Classifier Station consists of:
  • 20 Wastewater Classifier Strips
  • Two bobbins of polypropylene thread
  • Laminated Chemical Classifier Chart
  • One pair nitrile gloves
  • One plastic case
  • Order Info
    Additional test strips and classifier charts are available separately.
Specifications
Specifications
20 ppm fluoride, 10 ppm petroleum products/organic solvents, 10 ppm hydrogen sulfide, 10 ppm nitrates, 1 ppm nitrites
Classifying the wastewater for proper treatment
Each
Wastewater Classifier Kit
Six 11.75 L x 1 in. W wastewater classifier strips, one bobbin of polypropylene thread, laminated chemical classifier chart
0 to 13