missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

FyterTech Nonwovens Spilfyter™ Wastewater Classifiers

Distinguish risks of unknown spilled liquids

$284.20 - $939.37

Specifications

For Use With (Application) Classifying the wastewater for proper treatment
pH Range 0 to 13
View More Specs

Products 3
Catalog Number Mfr. No. Type Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Type Packaging Quantity Price Quantity  
19-065-574
View Documents
FyterTech Nonwovens
580010
Wastewater Classifier Test Strip 10/Pk.
Pack of 10 Each for $284.20
 
19-120-875
View Documents
FyterTech Nonwovens
580001
Wastewater Classifier Kit Each
Each for $284.20
 
19-812-534
View Documents
FyterTech Nonwovens
580050
Wastewater Classifier Test Strip 50/Pk.
Case of 50 Each for $939.37
 
Description

Description

Classifier Strips show responder if unknown spill contains critical levels of these contaminant categories: acid/base; petroleum/organic solvent; hydrogen sulfide; nitrite; nitrate; fluoride. Classifier chart compares standard colors to sample colors.

Wastewater Classifier Kit consists of:

  • Six Wastewater Classifier Strips
  • One bobbin of polypropylene thread
  • Laminated Chemical Classifier Chart
Wastewater Classifier Station consists of:
  • 20 Wastewater Classifier Strips
  • Two bobbins of polypropylene thread
  • Laminated Chemical Classifier Chart
  • One pair nitrile gloves
  • One plastic case
  • Order Info

    Additional test strips and classifier charts are available separately.

Specifications

Specifications

Classifying the wastewater for proper treatment
0 to 13
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'