Cytiva Spin Adapters for Gravity Columns
Convenient and rapid cleanup with high reproducibility and the potential for high throughput by running multiple samples in parallel, High desalting capacity, The use of centrifugation protocol with all gravity columns.
Supplier:  Cytiva 28923245
 View more versions of this product
Catalog No. 45-001-552
Explore available promotions
Description
Description
  • Convenient adapters, enable spinning of all gravity columns.
Specifications
Specifications
Convenient and rapid cleanup with high reproducibility and the potential for high throughput by running multiple samples in parallel, High desalting capacity, The use of centrifugation protocol with all gravity columns.
10 Pack
Spin Column Adapter
Polypropylene
Convenient and rapid cleanup with high reproducibility for high throughput by running multiple samples in PD-10 desalting column for centrifugation