Cytiva Spin Adapters for Gravity Columns
Convenient and rapid cleanup with high reproducibility and the potential for high throughput by running multiple samples in parallel, High desalting capacity, The use of centrifugation protocol with all gravity columns.
$27.57 - $36.64
Specifications
Description Convenient and rapid cleanup with high reproducibility and the potential for high throughput by running multiple samples in parallel, High desalting capacity, The use of centrifugation protocol with all gravity columns.
Material Polypropylene
Quantity 10 Pack
Type Spin Column Adapter
View More Specs
Products 3
Catalog Number Mfr. No. Type For Use With (Application) Price Quantity  
Catalog Number Mfr. No. Type For Use With (Application) Price Quantity  
45-001-552
View Documents
Cytiva
28923245
Spin Column Adapter Convenient and rapid cleanup with high reproducibility for high throughput by running multiple samples in PD-10 desalting column for centrifugation
Pack of 10 Each for $36.64
45-001-551
View Documents
Cytiva
28923244
Spin Column Adapter Convenient and rapid cleanup with high reproducibility for high throughput by running multiple samples in PD MidiTrap G-25 column for centrifugation
Pack of 10 Each for $36.63
45002004
View Documents
Cytiva
28923243
Spin Column Adapter Convenient and rapid cleanup with high reproducibility for high throughput by running multiple samples in PD MiniTrap G-25 column for centrifugation
Pack of 10 Each for $27.57
Description
Description
  • Convenient adapters, enable spinning of all gravity columns.
Specifications
Specifications
Convenient and rapid cleanup with high reproducibility and the potential for high throughput by running multiple samples in parallel, High desalting capacity, The use of centrifugation protocol with all gravity columns.
Polypropylene
10 Pack
Spin Column Adapter
SDS
missing translation for 'documents'
missing translation for 'documents'