Corning™ Square and Rectangular Cover Glasses
Provides quality, reliability and performance for your laboratory microscopy needs
$235.09 - $1029.70
Specifications
Product Type | Coverslip |
---|---|
Material | Borosilicate Glass |
Sterility | Non-sterile |
For Use With (Equipment) | Microscopes |
Catalog Number | Mfr. No. | Thickness (Metric) | Length (Metric) | Width (Metric) | Shape | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Thickness (Metric) | Length (Metric) | Width (Metric) | Shape | Price | Quantity | ||
12-553-452
|
Corning™
284525 |
#1 (0.13 to 0.16 mm) | 25 mm | 25 mm | Square |
Case of 2000 Each for $740.02
|
|
||
12-553-468
|
Corning™
2980225 |
#1.5 (0.16 to 0.19 mm) | 50 mm | 22 mm | Rectangle |
Case of 1000 Each for $774.18
|
|
||
12-553-455
|
Corning™
285025 |
#1.5 (0.16 to 0.19 mm) | 25 mm | 25 mm | Square |
Case of 2000 Each for $893.93
|
|
||
12-553-463
|
Corning™
2975244 |
#1 (0.13 to 0.16 mm) | 40 mm | 24 mm | Rectangle |
Case of 1000 Each for $566.43
|
|
||
12-553-460
|
Corning™
2975224 |
#1 (0.13 to 0.16 mm) | 40 mm | 22 mm | Rectangle |
Case of 1000 Each for $504.59
|
|
||
12-553-461
|
Corning™
2975225 |
#1 (0.13 to 0.16 mm) | 50 mm | 22 mm | Rectangle |
Case of 1000 Each for $637.25
|
|
||
12-553-458
|
Corning™
285525 |
#2 (0.19 to 0.23 mm) | 25 mm | 25 mm | Square |
Case of 1000 Each for $455.14
|
|
||
12-553-470
|
Corning™
2980244 |
#1.5 (0.16 to 0.19 mm) | 40 mm | 24 mm | Rectangle |
Case of 1000 Each for $701.73
|
|
||
12-553-464
|
Corning™
2975245 |
#1 (0.13 to 0.16 mm) | 50 mm | 24 mm | Rectangle |
Case of 1000 Each for $702.77
|
|
||
12-553-462
|
Corning™
2975243 |
#1 (0.13 to 0.16 mm) | 30 mm | 24 mm | Rectangle |
Case of 1000 Each for $423.00
|
|
||
12-553-471
|
Corning™
2980245 |
#1.5 (0.16 to 0.19 mm) | 50 mm | 24 mm | Rectangle |
Case of 1000 Each for $831.61
|
|
||
12-553-472
|
Corning™
2980246 |
#1.5 (0.16 to 0.19 mm) | 60 mm | 24 mm | Rectangle |
Case of 1000 Each for $1,029.70
|
|
||
12-553-466
|
Corning™
2980223 |
#1.5 (0.16 to 0.19 mm) | 30 mm | 22 mm | Rectangle |
Case of 1000 Each for $462.93
|
|
||
12-553-454
|
Corning™
285022 |
#1.5 (0.16 to 0.19 mm) | 22 mm | 22 mm | Square |
Case of 2000 Each for $694.50
|
|
||
12-553-457
|
Corning™
285522 |
0.17 to 0.25 mm | 22 mm | 22 mm | Square |
Case of 1000 Each for $354.39
|
|
||
12-553-465
|
Corning™
2975246 |
#1 (0.13 to 0.16 mm) | 60 mm | 24 mm | Rectangle |
Case of 1000 Each for $849.50
|
|
||
12-553-456
|
Corning™
285518 |
0.17 to 0.25 mm | 18 mm | 18 mm | Square |
Case of 1000 Each for $235.09
|
|
||
12-553-469
|
Corning™
2980243 |
#1.5 (0.16 to 0.19 mm) | 30 mm | 24 mm | Rectangle |
Case of 1000 Each for $605.43
|
|
||
12-553-453
|
Corning™
285018 |
#1.5 (0.16 to 0.19 mm) | 18 mm | 18 mm | Square |
Case of 2000 Each for $475.68
|
|
||
12-553-450
|
Corning™
284518 |
#1 (0.13 to 0.16 mm) | 18 mm | 18 mm | Square |
Case of 2000 Each for $382.39
|
|
||
12-553-451
|
Corning™
284522 |
#1 (0.13 to 0.16 mm) | 22 mm | 22 mm | Square |
Case of 2000 Each for $573.48
|
|
||
12-553-467
|
Corning™
2980224 |
#1.5 (0.16 to 0.19 mm) | 40 mm | 22 mm | Rectangle |
Case of 1000 Each for $673.68
|
|
||
Description
Provides quality, reliability and performance for your laboratory microscopy needs
- Made from soda-lime glass and borosilicate glass with high clarity
- Available in square and rectangular shapes, and in three standard sizes and thicknesses
Order Info
Packaged in plastic boxes for protection during storage.
Specifications
Coverslip | |
Borosilicate Glass | |
Non-sterile | |
Microscopes |