METTLER TOLEDO Standard LA Analytical Balance
METTLER TOLEDO Standard LA Analytical Balances have a precise weigh cell and external adjustment. The bright, backlit LCD display is equipped with 3 built-in applications for piece counting and dynamic weighing.
Supplier: METTLER TOLEDO 30823718
Description
Designed with robust construction, the overload protection and metal base can withstand tough weighing environments.
LA Analytical Balances include:
- Adjustment with external weights
- Bright backlit LCD Display
- 3 built-in weighing applications
- Front level indicator
- Overload protection
- Metal base, ABS top housing
- RS232 interface
- Bluetooth option
- Transfer weight function for easy data transfer
See how it ranks among our balance lines:
- XPR Excellence Analytical Balances
- MX Advanced Analytical Balances
- MR Advanced Analytical Balances
- MA Standard Analytical Balances
- ME Standard Analytical Balances
- LA Standard Analytical Balances
Specifications
USB-A, RS232 | |
Backlit LCD | |
Aluminum, ABS | |
3 sec. | |
100/240 V, 50/60 Hz |
82 g | |
0.1 mg | |
LA84E | |
Analytical Balance | |
No |