missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Starplex™ Scientific StarCLICK 100 Specimen Container

StarCLICK 100 features an audible, double click to confirm the cap is applied correctly. Ideal for leakproof specimen transport.

$325.68 - $325.68

Specifications

Disposable Single-use
Latex Free Yes
Volume (Metric) 100 mL
View More Specs

Products 1
Catalog Number Mfr. No. Closure Color Label Description Sterility No. per Pack Price Quantity  
Catalog Number Mfr. No. Closure Color Label Description Sterility No. per Pack Price Quantity  
22-046-578
View Documents
Starplex™ Scientific
B1002YCCNLTS
Yellow No label Non-sterile 400 Pk.
Case of 400 Each for $325.68
 
Description

Description

  • Polypropylene
  • Leakproof transport
  • Clear graduations on two sides in ounces and milliliters
  • Wide mouth design
  • Stackable
  • Available in sterile and non-sterile format
  • Externally validated to meet 95 kPa and IATA standards
Specifications

Specifications

Single-use
Yes
100 mL
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'