Starplex™ Scientific Histoplex™ Histology Containers
Specifically engineered for the storage of histology specimens
Supplier:  Starplex™ Scientific H602FL
 View more versions of this product
Catalog No. 15-182-72
Explore available promotions
Description
Description
Specifically engineered for the storage of histology specimens
  • O-ring polyethylene screw cap forms a seal that reduces most formalin leakage and evaporation
  • Durable polypropylene containers are translucent, flexible, resist cracking and splitting, and eliminate problems associated with formalin evaporation
  • 40mL to 1L containers have graduations for easy fill line viewing
  • 2 to 8L containers are white polypropylene with unique tamper-evident snap-on lids
Specifications
Specifications
Container
Yes
Non-sterile
Yellow
Clear
Yes
Polypropylene
60 mL
Screw Cap with O-Ring
Affixed