Starplex™ Scientific Histoplex™ Histology Containers
Specifically engineered for the storage of histology specimens
$320.61 - $567.16
Catalog Number | Mfr. No. | Capacity (Metric) | Closure Type | Closure Color | Label | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Closure Type | Closure Color | Label | Price | Quantity | ||
15-182-71
|
Starplex™ Scientific
H402FL |
40 mL | Screw Cap with O-Ring | Yellow | In case |
Case of 600 Each for $399.58
|
|
||
15-182-72
|
Starplex™ Scientific
H602FL |
60 mL | Screw Cap with O-Ring | Yellow | Affixed |
Case of 500 Each for $521.55
|
|
||
15-182-73
|
Starplex™ Scientific
H902FL |
90 mL | Screw Cap with O-Ring | Yellow | Affixed |
Case of 400 Each for $458.92
|
|
||
15-182-78
|
Starplex™ Scientific
H50002 |
5 L | Snap Cap | White | In case |
Case of 10 Each for $320.61
|
|
||
15-182-77
|
Starplex™ Scientific
H20002 |
2 L | Snap Cap | White | In case |
Case of 20 Each for $356.55
|
|
||
15-182-79
|
Starplex™ Scientific
H80002 |
8 L | Snap Cap | White | In case |
Case of 10 Each for $352.68
|
|
||
15-182-80
|
Starplex™ Scientific
H5002C |
500 mL | Screw Cap with O-Ring | Yellow | In case |
Case of 100 Each for $460.58
|
|
||
15-182-84
|
Starplex™ Scientific
H2502C |
250 mL | Screw Cap with O-Ring | Yellow | In case |
Case of 100 Each for $356.21
|
|
||
15-182-85
|
Starplex™ Scientific
H10002C |
1 L | Screw Cap with O-Ring | Yellow | In case |
Case of 100 Each for $567.16
|
|
||
Description
Specifically engineered for the storage of histology specimens
- O-ring polyethylene screw cap forms a seal that reduces most formalin leakage and evaporation
- Durable polypropylene containers are translucent, flexible, resist cracking and splitting, and eliminate problems associated with formalin evaporation
- 40mL to 1L containers have graduations for easy fill line viewing
- 2 to 8L containers are white polypropylene with unique tamper-evident snap-on lids
Specifications
Yes | |
Polypropylene | |
Non-sterile |