missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Stearic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 174490010

 View more versions of this product

Catalog No. AC174490010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Stearic acid
67.0°C to 69.0°C
0.8400g/mL
196°C
96% min. (GC)
C18H36O2
MFCD00002752
02,IV,1206
15,893
>90 °C
QIQXTHQIDYTFRH-UHFFFAOYSA-N
octadecanoic acid
5281
284.47
Granular Powder, Granules, or Flakes
57-11-4
White
361.0°C
Authentic
Plastic Bottle
CH3(CH2)16CO2H
1 kg
0.84
stearic acid, stearophanic acid, n-octadecanoic acid, cetylacetic acid, pearl stearic, stearex beads, octadecansaeure, stearinsaeure, vanicol, 1-heptadecanecarboxylic acid
Solubility in water: slightly soluble. Other solubilities: 1g/21mL alcohol- 1g/6mL ccl4,1g/5mL benzene- 1g/3.4mL cs2,1g/2mL chloroform - 1g/26mL acetone,soluble in amyl acetate and toluene
CCCCCCCCCCCCCCCCCC(O)=O
284.48
CHEBI:28842
97%

Chemical Identifiers

57-11-4
284.48
QIQXTHQIDYTFRH-UHFFFAOYSA-N
5281
octadecanoic acid
C18H36O2
MFCD00002752
stearic acid, stearophanic acid, n-octadecanoic acid, cetylacetic acid, pearl stearic, stearex beads, octadecansaeure, stearinsaeure, vanicol, 1-heptadecanecarboxylic acid
CHEBI:28842
CCCCCCCCCCCCCCCCCC(O)=O
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-313-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only